4-tert-Butyl-1-bromobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromo-4-tert-butylbenzene, 2-(4-Bromophenyl)-2-methylpropane, 1-Bromo-4-(tert-butyl)benzene, 4-Bromo-tert-butylbenzene
A133502-100G
A133502-10G
A133502-500G
A133502-1000G
A133502-25G
A133502-5G
103808-462EA
143
USD
103808-462
77095-078
77095-080
77095-082
77314-534
77314-536
77314-538
4-tert-Butyl-1-bromobenzene 98%
4-tert-Butyl-1-bromobenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₃Br MW: 213.12 g/mol Boiling Pt: 232 °C (760 mm Hg) Melting Pt: 13…16 °C Flash Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000108 CAS Number: 3972-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...