1-Bromo-4-(tert-butyl)-2-chlorobenzene 96%

Supplier: Ambeed
Warning

A123027-250MG A123027-25G A123027-1G A123027-5G
103800-330EA 13 USD
103800-330 76870-322 76870-324 77297-916 77297-918
1-Bromo-4-(tert-butyl)-2-chlorobenzene 96%
1-Bromo-4-(tert-butyl)-2-chlorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrCl
MW: 247.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072806
CAS Number: 1251032-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR