1-Benzyloxy-4-bromobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyloxybromobenzene, 1-Bromo-4-(phenylmethoxy)benzene

A131440-100G A131440-10G A131440-500G A131440-5G A131440-25G
77285-336EA 44.29 USD
77285-336 77285-338 77285-340 77298-482 77314-084
1-Benzyloxy-4-bromobenzene 98%
1-Benzyloxy-4-bromobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Boiling Pt: 340.6 °C (760 mm Hg)
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00028016
CAS Number: 6793-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR