1-Bromo-4-hexyloxybenzene 97%

Supplier: Ambeed
Warning

A101688-25G A101688-1G A101688-5G
103796-092EA 12 USD
103796-092 103821-784 76717-564 77282-800 77282-802
1-Bromo-4-hexyloxybenzene 97%
1-Bromo-4-hexyloxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrO
Boiling Pt: 298.4 °C (760 mm Hg)
Flash Pt: 126.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00173753
CAS Number: 30752-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR