4-(2,2,2-Trifluoroethoxy)bromobenzene 97%
Supplier: Ambeed
|
Warning
|
A115433-10G
A115433-25G
A115433-1G
103798-562EA
24
USD
103798-562
103824-418
76869-982
76869-984
77297-370
4-(2,2,2-Trifluoroethoxy)bromobenzene 97%
4-(2,2,2-Trifluoroethoxy)bromobenzene
Precaución: for Research Use Only
Formula:
C₈H₆BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD07780193 CAS Number: 106854-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...