3-Bromostyrene 95% stabilized

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-vinylbenzene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A267486-1G A267486-25G A267486-5G A267486-100G A267486-10G
103812-856EA 6 USD
103812-856 103852-756 77092-498 77092-500 77092-502 77098-954 77098-956
3-Bromostyrene 95% stabilized
3-Bromostyrene

Precaución: for Research Use Only

Formula: C₈H₇Br
MW: 183.05 g/mol
Boiling Pt: 227.6 °C (760 mm Hg)
Flash Pt: 86.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000088
CAS Number: 2039-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR