3-Bromophenyl phenyl ether (BDE No. 2) 98%

Supplier: Ambeed
Danger

Synonyms: 3-Bromodiphenyl ether, 1-Bromo-3-phenoxybenzene

A456205-1G A456205-5G A456205-10G A456205-250MG A456205-25G
103827-856EA 28 USD
103827-856 103838-930 76842-780 76842-782 77291-744 77291-746 77307-398
3-Bromophenyl phenyl ether (BDE No. 2) 98%
3-Bromophenyl phenyl ether (BDE No. 2)

Precaución: for Research Use Only

Formula: C₁₂H₉BrO
MW: 249.1000 g/mol
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00474520
CAS Number: 6876-00-2
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR