3-Bromo-5-nitrotoluene 98%

Supplier: Ambeed
Warning

A107034-100G A107034-10G A107034-25G A107034-1G A107034-5G
103797-116EA 11 USD
103797-116 103822-874 77135-722 77135-724 77135-726 77309-744 77309-746
3-Bromo-5-nitrotoluene 98%
3-Bromo-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
Boiling Pt: 269.5 °C (760 mm Hg)
Melting Pt: 68…72 °C
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD06659815
CAS Number: 52488-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR