3-Bromo-5-methylbenzotrifluoride 98%
Supplier: Ambeed
|
Warning
|
A230321-10G
A230321-250MG
A230321-1G
A230321-25G
A230321-5G
A230321-100G
103819-538EA
9
USD
103819-538
103850-436
76786-244
76786-246
77089-294
77303-870
77303-872
77872-168
3-Bromo-5-methylbenzotrifluoride 98%
3-Bromo-5-methylbenzotrifluoride
Precaución: for Research Use Only
Formula:
C₈H₆BrF₃ Boiling Pt: 178.7 °C (760 mm Hg) Flash Pt: 67.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631585 CAS Number: 86845-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...