3-Bromo-2-nitrotoluene 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromo-3-methyl-2-nitrobenzene
A194155-10G
A194155-1G
A194155-25G
A194155-5G
103816-624EA
6
USD
103816-624
103847-234
76769-952
77085-150
77085-152
77085-154
3-Bromo-2-nitrotoluene 97%
3-Bromo-2-nitrotoluene
Precaución: for Research Use Only
Formula:
C₇H₆BrNO₂ MW: 216.03 g/mol Boiling Pt: 237.4 °C (760 mm Hg) Melting Pt: 26.8…29 °C Flash Pt: 97.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD02673631 CAS Number: 52414-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...