3-Bromoanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-methoxybenzene

A135295-100G A135295-10G A135295-500G A135295-1000G A135295-25G
103808-502EA 103 USD
103808-502 77109-486 77109-488 77109-490 77314-934 77314-936
3-Bromoanisole 98%
3-Bromoanisole

Precaución: for Research Use Only

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 210.5 °C (760 mm Hg)
Melting Pt: 2 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00000081
CAS Number: 2398-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR