3-Bromo-5-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-methoxy-5-nitrobenzene

A141768-100G A141768-10G A141768-250MG A141768-500G A141768-1G A141768-25G A141768-5G
103803-056EA 6 USD
103803-056 77109-832 77109-834 77109-836 77109-838 77316-236 77316-238 77316-240
3-Bromo-5-nitroanisole 98%
3-Bromo-5-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.0300 g/mol
Boiling Pt: 295.3 °C (760 mm Hg)
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD04112752
CAS Number: 16618-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR