3-Bromo-5-methylanisole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-5-methoxytoluene, 1-Bromo-3-methoxy-5-methylbenzene
A131087-100G
A131087-1G
A131087-25G
A131087-5G
103801-520EA
9
USD
103801-520
77246-536
77313-996
77313-998
77314-000
3-Bromo-5-methylanisole 97%
3-Bromo-5-methylanisole
Precaución: for Research Use Only
Formula:
C₈H₉BrO MW: 201.0600 g/mol Boiling Pt: 228.4 °C (760 mm Hg) Flash Pt: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD08061916 CAS Number: 29578-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...