3-Bromo-5-methoxybenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-methoxy-5-(trifluoromethyl)benzene, 1-Bromo-3-(trifluoromethyl)-5-methoxybenzene

A106828-100G A106828-10G A106828-25G A106828-5G A106828-1G
103797-084EA 14 USD
103797-084 103822-842 76869-476 76869-478 76869-480 77283-518 77309-696
3-Bromo-5-methoxybenzotrifluoride 98%
3-Bromo-5-methoxybenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999639
CAS Number: 627527-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR