1-Bromo-3-fluoro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A146070-100G A146070-10G A146070-1G A146070-25G A146070-5G
103803-626EA 10 USD
103803-626 103841-712 77110-056 77110-058 77317-014 77317-016 77317-018
1-Bromo-3-fluoro-5-nitrobenzene 98%
1-Bromo-3-fluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
Boiling Pt: 245.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09955454
CAS Number: 7087-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR