3-Bromo-α,α,α,5-tetrafluorotoluene 95%

Supplier: Ambeed
Warning

A145970-100G A145970-10G A145970-500G A145970-5G A145970-25G
103841-694EA 5 USD
103841-694 76767-986 76767-988 76767-990 77299-484 77317-002
3-Bromo-α,α,α,5-tetrafluorotoluene 95%
3-Bromo-α,α,α,5-tetrafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 151 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00042498
CAS Number: 130723-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR