1-Bromo-3-fluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A134027-100G A134027-10G A134027-500G A134027-1G A134027-25G A134027-5G
103801-958EA 9 USD
103801-958 77109-416 77109-418 77109-420 77314-668 77314-670 77314-672
1-Bromo-3-fluoro-2-nitrobenzene 98%
1-Bromo-3-fluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
Boiling Pt: 232 °C (760 mm Hg)
Melting Pt: 36…37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368788
CAS Number: 886762-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR