1-Bromo-3-fluoro-2-iodobenzene 97%
Supplier: Ambeed
|
Warning
|
A265408-1G
A265408-25G
A265408-5G
A265408-10G
103812-728EA
6
USD
103812-728
103852-622
77092-306
77092-308
77092-310
77672-374
1-Bromo-3-fluoro-2-iodobenzene 97%
1-Bromo-3-fluoro-2-iodobenzene
Precaución: for Research Use Only
Formula:
C₆H₃BrFI Boiling Pt: 248.6 °C (760 mm Hg) Flash Pt: 104.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368675 CAS Number: 450412-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...