1-Bromo-3-ethoxy-5-fluorobenzene 98%

Supplier: Ambeed
Warning

A134534-10G A134534-25G
103802-020EA 11 USD
103802-020 76823-160 77314-780
1-Bromo-3-ethoxy-5-fluorobenzene 98%
1-Bromo-3-ethoxy-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.05 g/mol
Boiling Pt: 207.2±20.0 °C (760 mm Hg)
Flash Pt: 96.4 °C
Storage Temperature: Ambient
MDL Number: MFCD18825150
CAS Number: 212307-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR