3-Bromo-5-chloroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-chloro-5-methoxybenzene

A143484-100G A143484-10G A143484-250MG A143484-1G A143484-25G A143484-5G
103803-288EA 7 USD
103803-288 76783-026 76783-028 76783-030 77316-558 77316-560 77316-562
3-Bromo-5-chloroanisole 97%
3-Bromo-5-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.48 g/mol
Boiling Pt: 236.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11035921
CAS Number: 174913-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR