3-Bromo-5-chlorobenzotrifluoride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromo-3-chloro-5-(trifluoromethyl)benzene
A401542-25G
A401542-5G
A401542-100G
A401542-10G
A401542-1G
A401542-500G
103838-094EA
8
USD
103838-094
76841-556
76841-558
76979-064
76979-066
76979-068
76979-070
3-Bromo-5-chlorobenzotrifluoride 98%
3-Bromo-5-chlorobenzotrifluoride
Precaución: For research use only
Formula:
C₇H₃BrClF₃ MW: 259.45 g/mol Boiling Pt: 182 °C (760 mm Hg) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD09260894 CAS Number: 928783-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...