1-Bromo-3-chloro-2-fluorobenzene 98%

Supplier: Ambeed
Warning

A126022-100G A126022-10G A126022-1G A126022-500G A126022-25G A126022-5G
77285-112EA 48 USD
77285-112 77285-114 77285-116 77285-118 77312-918 77312-920
1-Bromo-3-chloro-2-fluorobenzene 98%
1-Bromo-3-chloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClF
Boiling Pt: 197.9 °C (760 mm Hg)
Melting Pt: 44 °C
Flash Pt: 73.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03412182
CAS Number: 144584-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR