2-Bromo-6-chlorobenzotrifluoride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-chloro-2-(trifluoromethyl)benzene

A203887-1G A203887-5G A203887-100G A203887-10G A203887-25G
103817-504EA 9 USD
103817-504 103848-164 77086-306 77086-308 77139-420 77139-422 77302-880
2-Bromo-6-chlorobenzotrifluoride 97%
2-Bromo-6-chlorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃
MW: 259.4500 g/mol
Boiling Pt: 210.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08459291
CAS Number: 857061-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR