1-Bromo-3,5-di-tert-butylbenzene 98%

Supplier: Ambeed
Warning

A194017-25G A194017-5G A194017-100G A194017-10G A194017-500G A194017-1G
103816-612EA 6 USD
103816-612 103847-220 77085-130 77085-132 77111-896 77111-898 77111-900 77302-380
1-Bromo-3,5-di-tert-butylbenzene 98%
1-Bromo-3,5-di-tert-butylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₂₁Br
MW: 269.23 g/mol
Melting Pt: 62…66 °C
Flash Pt: 98.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00796945
CAS Number: 22385-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR