1-Bromo-3,5-dimethoxybenzene 97%

Supplier: Ambeed
Warning

A105389-100G A105389-10G A105389-1G A105389-25G A105389-5G A105389-1000G A105389-500G
103822-612EA 6 USD
103822-612 77107-538 77107-540 77107-542 77107-544 77309-484 77549-004 77549-006
1-Bromo-3,5-dimethoxybenzene 97%
1-Bromo-3,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
Boiling Pt: 246 °C (760 mm Hg)
Melting Pt: 62…66 °C
Flash Pt: 103.1 °C
Storage Temperature: Ambient
MDL Number: MFCD06200685
CAS Number: 20469-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR