1-Bromo-3,5-difluoro-2-iodobenzene 97%

Supplier: Ambeed
Warning

A172305-1G A172305-25G A172305-5G A172305-100G A172305-10G A172305-500G
103806-714EA 9 USD
103806-714 103844-998 77082-190 77082-192 77082-194 77151-318 77151-320 77151-322
1-Bromo-3,5-difluoro-2-iodobenzene 97%
1-Bromo-3,5-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂I
MW: 318.89 g/mol
Boiling Pt: 235.4 °C (760 mm Hg)
Melting Pt: 48…51 °C
Flash Pt: 96.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00042183
CAS Number: 175278-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR