3-tert-Butyl-1-bromobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-tert-butylbenzene, 1-Bromo-3-tert-butylbenzene, 1-Bromo-3-(tert-butyl)benzene, 2-(3-Bromophenyl)-2-methylpropane

A297883-1G A297883-25G A297883-5G A297883-10G A297883-100G
103814-156EA 9 USD
103814-156 103835-300 76837-986 76837-988 76837-990 77239-198 77550-804
3-tert-Butyl-1-bromobenzene 98%
3-tert-Butyl-1-bromobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 212.4 °C (760 mm Hg)
Flash Pt: 81.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00234661
CAS Number: 3972-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR