1-Bromo-3-(difluoromethyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-(difluoromethyl)benzene, 3-Bromobenzal fluoride

A125491-25G A125491-1G A125491-5G
103800-686EA 13 USD
103800-686 76781-964 77312-832 77312-834
1-Bromo-3-(difluoromethyl)benzene 98%
1-Bromo-3-(difluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.015 g/mol
Boiling Pt: 197.7 °C (760 mm Hg)
Flash Pt: 73.4 °C
Storage Temperature: Ambient
MDL Number: MFCD07368984
CAS Number: 29848-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR