3-Bromobenzyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-α-chlorotoluene, 1-(Chloromethyl)-3-bromobenzene, α-Chloro-3-bromotoluene, 1-Bromo-3-(chloromethyl)benzene

A167584-1G A167584-25G A167584-5G A167584-10G
103806-200EA 9 USD
103806-200 103844-460 77081-476 77081-478 77081-480 77273-586
3-Bromobenzyl chloride 98%
3-Bromobenzyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrCl
MW: 205.48 g/mol
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040865
CAS Number: 932-77-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR