4-Bromo-3-nitrobenzotrifluoride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-3-nitrobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-3-nitrotoluene, 1-Bromo-2-nitro-4-(trifluoromethyl)benzene
A204629-100G
A204629-10G
A204629-1G
A204629-25G
A204629-5G
103848-226EA
8
USD
103848-226
76770-282
76770-284
76770-286
77086-400
77086-402
4-Bromo-3-nitrobenzotrifluoride 98%
4-Bromo-3-nitrobenzotrifluoride
Precaución: for Research Use Only
Formula:
C₇H₃BrF₃NO₂ MW: 270.01 g/mol Boiling Pt: 225.9 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014685 CAS Number: 349-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...