1-Bromo-2-naphthaldehyde 98%

Supplier: Ambeed
Warning

A108898-10G A108898-25G A108898-1G A108898-5G
103797-398EA 14 USD
103797-398 103823-188 77233-392 77233-394 77310-016 77310-018
1-Bromo-2-naphthaldehyde 98%
1-Bromo-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₇BrO
MW: 235.0767 g/mol
Boiling Pt: 347.3 °C (760 mm Hg)
Flash Pt: 119.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00046368
CAS Number: 3378-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR