1-Bromo-2-methylnaphthalene 90%

Supplier: Ambeed
Warning

A296134-100G A296134-10G A296134-500G A296134-25G A296134-5G
103835-222EA 8 USD
103835-222 77239-170 77239-172 77239-174 77305-684 77305-686
1-Bromo-2-methylnaphthalene 90%
1-Bromo-2-methylnaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉Br
Boiling Pt: 300.9 °C (760 mm Hg)
Flash Pt: 137.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00003871
CAS Number: 2586-62-1
ADR: N/A,N/A

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR