1-Bromo-2-methoxynaphthalene 98%

Supplier: Ambeed
Warning

A147971-100G A147971-10G A147971-500G A147971-25G A147971-5G
103841-972EA 5 USD
103841-972 76798-518 76798-520 76798-522 77299-622 77317-396
1-Bromo-2-methoxynaphthalene 98%
1-Bromo-2-methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉BrO
Boiling Pt: 319.2 °C (760 mm Hg)
Melting Pt: 80…83 °C
Flash Pt: 132.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00055374
CAS Number: 3401-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR