2-Methoxy-4-(trifluoromethoxy)bromobenzene 96%

Supplier: Ambeed
Warning

A192679-100MG A192679-250MG A192679-1G A192679-5G
103816-474EA 22 USD
103816-474 103847-084 76825-318 76825-320 77084-958 77084-960
2-Methoxy-4-(trifluoromethoxy)bromobenzene 96%
2-Methoxy-4-(trifluoromethoxy)bromobenzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD08458014
CAS Number: 848360-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR