2-Bromo-6-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-methoxy-3-nitrobenzene

A242600-1G A242600-25G A242600-5G A242600-100G A242600-10G A242600-250MG
103811-480EA 6 USD
103811-480 103851-296 77090-478 77090-480 77090-482 77238-128 77238-130 77238-132
2-Bromo-6-nitroanisole 98%
2-Bromo-6-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
Boiling Pt: 296.4 °C (760 mm Hg)
Flash Pt: 133.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09261208
CAS Number: 98775-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR