1-Bromo-2-iodobenzene 98% stabilized

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-iodobenzene, 2-Bromoiodobenzene

Stabilization: Stabilized with Copper chip

A164868-100G A164868-10G A164868-250MG A164868-500G A164868-1000G A164868-25G A164868-5G
103808-950EA 314 USD
103808-950 103844-176 77110-814 77110-816 77110-818 77110-820 77300-740 77320-372 77320-374
1-Bromo-2-iodobenzene 98% stabilized
1-Bromo-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₄BrI
MW: 282.91 g/mol
Melting Pt: 9…10 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001030
CAS Number: 583-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR