4-Bromo-3-iodotoluene 98%

Supplier: Ambeed
Danger

A356951-25G A356951-5G A356951-1G
103826-180EA 5 USD
103826-180 103837-186 76840-332 76840-334 77306-684
4-Bromo-3-iodotoluene 98%
4-Bromo-3-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 271.5 °C (760 mm Hg)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD07784420
CAS Number: 858841-53-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR