4-Bromo-3-iodobenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-iodobenzotrifluoride, 1-Bromo-2-iodo-4-(trifluoromethyl)benzene

A407159-1G A407159-10G
103827-154EA 7 USD
103827-154 103838-194 76841-688 76979-122
4-Bromo-3-iodobenzotrifluoride 98%
4-Bromo-3-iodobenzotrifluoride

Precaución: For research use only

Formula: C₇H₃BrF₃I
MW: 350.9 g/mol
Boiling Pt: 262 °C (760 mm Hg)
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09800691
CAS Number: 640280-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR