3-Bromo-2-iodoanisole 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-iodo-3-methoxybenzene, 3-Bromo-2-iodophenyl methyl ether

A114096-10G A114096-25G A114096-1G
103798-346EA 26 USD
103798-346 103824-188 76971-068 76971-070 77311-012
3-Bromo-2-iodoanisole 95%
3-Bromo-2-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
MW: 312.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089327
CAS Number: 450412-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR