4-Bromo-α,α,α,3-tetrafluorotoluene 98%

Supplier: Ambeed
Warning

A170065-1G A170065-5G
103806-442EA 5 USD
103806-442 103844-708 77081-808 77081-810
4-Bromo-α,α,α,3-tetrafluorotoluene 98%
4-Bromo-α,α,α,3-tetrafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 174.6 °C (760 mm Hg)
Flash Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00042497
CAS Number: 40161-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR