1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A175622-1G A175622-25G A175622-5G
103807-068EA 7 USD
103807-068 103845-380 77301-388 77301-390 77301-392
1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene 98%
1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄O
MW: 259.00 g/mol
Boiling Pt: 158.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06657548
CAS Number: 168971-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR