1-Bromo-2-ethylbenzene 97%

Supplier: Ambeed
Warning

A101465-100G A101465-10G A101465-25G A101465-5G
103821-670EA 5 USD
103821-670 77245-124 77245-126 77245-128 77297-112
1-Bromo-2-ethylbenzene 97%
1-Bromo-2-ethylbenzene

Precaución: for Research Use Only

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 199 °C (760 mm Hg)
Melting Pt: …56 °C
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00000077
CAS Number: 1973-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR