(2-Bromoethyl)cyclohexane 95%

Supplier: Ambeed
Warning

Synonyms: 2-Cyclohexylethyl bromide, 1-Bromo-2-cyclohexylethane

A721287-1G A721287-25G A721287-5G
103840-750EA 22 USD
103840-750 76844-966 76844-968 76844-970
(2-Bromoethyl)cyclohexane 95%
(2-Bromoethyl)cyclohexane

Precaución: for Research Use Only

Formula: C₈H₁₅Br
MW: 191.11 g/mol
Boiling Pt: 212 °C (760 mm Hg)
Melting Pt: 57 °C (760 mm Hg)
Flash Pt: 81.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00019398
CAS Number: 1647-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR