2-Bromo-3-chloroiodobenzene 95%

Supplier: Ambeed
Warning

A115818-10G A115818-250MG A115818-25G A115818-1G A115818-5G
103798-626EA 12 USD
103798-626 103824-486 77136-258 77136-260 77136-262 77311-310 77311-312
2-Bromo-3-chloroiodobenzene 95%
2-Bromo-3-chloroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClI
Boiling Pt: 289.3±20.0 °C (760 mm Hg)
Flash Pt: 128.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD28397823
CAS Number: 1369793-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR