1-Bromo-2-chloro-4-methyl-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A163716-250MG A163716-5G A163716-1G
103805-756EA 28 USD
103805-756 103844-018 76824-182 77300-670 77320-128
1-Bromo-2-chloro-4-methyl-5-nitrobenzene 98%
1-Bromo-2-chloro-4-methyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059082
CAS Number: 1126367-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR