1-Bromo-2-chloro-4-iodobenzene 98%

Supplier: Ambeed
Warning

A123329-10G A123329-1G A123329-5G A123329-100G A123329-25G
103800-390EA 9 USD
103800-390 77026-600 77312-504 77312-506 77599-802 77666-766
1-Bromo-2-chloro-4-iodobenzene 98%
1-Bromo-2-chloro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrCl
Storage Temperature: Ambient
MDL Number: MFCD04973904
CAS Number: 535934-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR