1-Bromo-2-chloro-3-fluorobenzene 98%
Supplier: Ambeed
|
Warning
|
A145891-100G
A145891-10G
A145891-1G
A145891-500G
A145891-25G
A145891-5G
103841-686EA
11
USD
103841-686
77110-038
77110-040
77110-042
77110-044
77299-480
77299-482
1-Bromo-2-chloro-3-fluorobenzene 98%
1-Bromo-2-chloro-3-fluorobenzene
Precaución: for Research Use Only
Formula:
C₆H₃BrClF Boiling Pt: 205.7 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD04038306 CAS Number: 883499-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...