1-Bromo-2-chloro-3,5-difluorobenzene 98%

Supplier: Ambeed
Warning

A129304-10G A129304-1G A129304-5G A129304-250MG
103801-262EA 12 USD
103801-262 76983-590 77313-644 77313-646 77889-888
1-Bromo-2-chloro-3,5-difluorobenzene 98%
1-Bromo-2-chloro-3,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
Boiling Pt: 197.2±35.0 °C (760 mm Hg)
Flash Pt: 73.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00135441
CAS Number: 187929-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR