1-Bromo-2,5-dimethoxy-4-iodobenzene 95%
Supplier: Ambeed
|
Warning
|
A210207-10G
A210207-1G
A210207-5G
103817-986EA
77
USD
103817-986
103848-678
77112-424
77303-116
77303-118
1-Bromo-2,5-dimethoxy-4-iodobenzene 95%
1-Bromo-2,5-dimethoxy-4-iodobenzene
Precaución: for Research Use Only
Formula:
C₈H₈BrIO₂ MW: 342.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27665621 CAS Number: 174518-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...