2-Bromo-1,4-difluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Difluorobromobenzene, 1-Bromo-2,5-difluorobenzene

A357950-25G A357950-100G A357950-10G A357950-500G A357950-5G A357950-1000G
103837-206EA 5 USD
103837-206 76840-366 77101-154 77101-156 77106-880 77306-688 77613-036
2-Bromo-1,4-difluorobenzene 98%
2-Bromo-1,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 155.4 °C (760 mm Hg)
Melting Pt: …31 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000345
CAS Number: 399-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR